Learn More
Ubpbio Containing the fluorogenic substrate Ub-Rhodamine 110 and 10 DUBs with representative members in each main DUB category. DUB activity validated using the included optimized buffer
Supplier:  Ubpbio J6110
Encompass_Preferred
This kit is designed to determine DUB inhibitor specificity and potency. You can select 10 DUBs of your choice from our DUB list. Each DUB is sufficient for 20 X 50ul reactions using a working concentration of 10nM. The supplied Ub-rhodamine 110 is sufficient for 550 X 50ul reactions using a working concentration of 100nM. The supplied iodoacetamide can be used as an inhibitor of cysteine DUBs using a working concentration of 10mM.
Catalog No. 50-233-9301
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.