Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deuterium bromide, 48% w/w in D{2}O, 99+% (Isotopic)

Catalog No. AA4240809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4240809 10 g
AA4240818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4240809 Supplier Thermo Scientific Chemicals Supplier No. 04240809
Add to Cart

NMR Solvent | CAS: 13536-59-9 | BrH | 81.918 g/mol

NMR Solvent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13536-59-9
Molecular Formula BrH
Molecular Weight (g/mol) 81.918
MDL Number MFCD00044232
InChI Key CPELXLSAUQHCOX-DYCDLGHISA-N
Synonym deuterium bromide, hydrobromic acid-d, deutero-bromide, dbr solution, deuterium bromide solution, bromo 2 h, deuterium bromide, 99 atom % d, deuterium bromide w/w in d2o, deuterium bromide 98.0atom%d in d2o, deuterium bromide solution, 47 wt. % in d2o, 99 atom % d
PubChem CID 518779
SMILES Br

Specifications

Physical Form Liquid
Quantity 10 g
Color Colorless
Packaging Clear Glass Ampule
Sensitivity Light and Moisture
Boiling Point 126°C
UN Number UN1788
Formula Weight 81.93
Percent Purity ≥99%
Concentration or Composition (by Analyte or Components) 48% w/w in D2O
Chemical Name or Material Deuterium bromide, Isotopic
Show More Show Less
Hazard Category H314-H331-H335
Hazard Statement GHS H Statement
H314-H373-H332-H335
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROBROMIC ACID
EINECSNumber 236-894-6
TSCA No
storageNote1 2°C to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.