Learn More
Perkin Elmer US LLC Deuterium Lamp (PDA Plus)
Supplier:  Perkin Elmer US LLC 09290900
Encompass_Preferred
Some functional attributes include: Voltage: 90 V Voltage Type: Vdc Current [from]: 270 mA Current [to]: 330 mA Ignition Voltage: 2.5 V Ignition Voltage Type: Vdc Length: 59 mm Diameter: 30 mm Material: UV GLASS WINDOW Light Aperature: 0.5 mm
Catalog No. 50-210-5068
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.