Learn More
Deuterium oxide, deuteration degree min. 99.9% for NMR spectroscopy MagniSolv™, MilliporeSigma™
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Physical Form | Liquid |
| Quantity | 10 mL |
| Packaging | Septum Ampule |
| Color | Colorless |
| Odor | Odorless |
| Assay Percent Range | 99.90% |
| Formula Weight | 20.03 |
| Chemical Name or Material | Deuterium oxide |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.