Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deuterium oxide, deuteration degree min. 99.96% for NMR spectroscopy, MagniSolv™, MilliporeSigma™

Catalog No. M1034280010 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
10 x 0.5 mL
10 x 0.75 mL
Packaging:
Glass Ampule
Glass Bottle
Septum Ampule
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1034280010 10 mL Septum Ampule
M1034280005 10 x 0.5 mL Glass Ampule
M1034280100 100 mL Glass Bottle
M1034280009 10 x 0.75 mL Glass Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M1034280010 Supplier MilliporeSigma Supplier No. 1.03428.0010
Add to Cart
Add to Cart

For NMR Spectroscopy

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-20-0
Molecular Formula H2O
Molecular Weight (g/mol) 20.03
MDL Number MFCD00044636
InChI Key XLYOFNOQVPJJNP-ZSJDYOACSA-N
Synonym deuterium oxide, water-d2, heavy water, deuterated water, dideuterium oxide, heavy water-d2, heavy water d2o, water sup 2-h2, deuterium oxide usan, water, heavy d2-o
PubChem CID 24602
ChEBI CHEBI:41981
IUPAC Name [(²H)oxy](²H)
SMILES [2H]O[2H]

Specifications

Physical Form Liquid
Packaging Septum Ampule
Color Colorless
Quantity 10 mL
Odor Odorless
Assay Percent Range 99.96% min.
Formula Weight 20.03
Chemical Name or Material Deuterium oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.