Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Dexamethasone
SDP

Catalog No. TB1126RMU50
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TB1126RMU50 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. TB1126RMU50 Supplier Tocris Bioscience™ Supplier No. TB1126RMU/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dexamethasone synthesized to Ancillary Material Grade

TB1126-RMU is an Ancillary Material Grade version of Dexamethasone (Cat. No. 1126). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies.

Find out more about our Ancillary Material Grade product range.

Specifications

Chemical Name or Material (11β,16α)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione
CAS 50-02-2
Quantity 50 mg
Target Glucocorticoid Receptor Compounds
Molecular Formula C22H29FO5
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.