Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Dexamethasone, Anti-inflammatory Glucocorticoid, Solid, 392.5 Da, 100 mg
Description
- Purity: >99%
- Molecular Weight: 392.5 Da
- Biological Activity: Anti-inflammatory glucocorticoid (IC50 = 38 nM and Ki = 6.7 nM for the glucocorticoid receptor). Displays 7-fold higher affinity for glucocorticoid receptors than hydrocortisone.
- Applications: Used in the generation of liver organoids.
- Form: Solid
- Constituents: 100% Dexamethasone
- CAS Number: 50-02-2
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Glucocorticoid Receptor |
| Shipping Condition | Ambient |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Formulation | 100% Dexamethasone |
| Quantity | 100 mg |
| For Use With (Application) | Generation of liver organoids |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?