Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dexamethasone, MP Biomedicals™

Catalog No. ICN19004094 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19004094 5 g
ICN19004090 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19004094 Supplier MP Biomedicals, Inc Supplier No. 0219004094
Add to Cart
Add to Cart

Anti-inflammatory agent

Dexamethasone is a glucocorticoid anti-inflammatory agent. It regulates T cell survival, growth, and differentiation. It also inhibits the induction of nitric oxide synthase.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dexamethasone
CAS 50-02-2
Synonym maxidex
Quantity 5 g
Molecular Formula C22H29FO5
Formula Weight 392.47
Physical Form Powder
Color White
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.