Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dexamethasone, MP Biomedicals™

Catalog No. MP21993501 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21993501 100 mg
MP21993506 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP21993501 Supplier MP Biomedicals, Inc Supplier No. 02199350.1
Add to Cart
Add to Cart

Used to study apoptosis, signaling pathways and gene expression. Dexamethasone, MP Biomedicals is an anti-inflammatory glucocorticoid with a range of effects on cell survival, cell signaling and gene expression.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-02-2
Molecular Formula C22H29FO5
Molecular Weight (g/mol) 392.47
MDL Number MFCD00064136
InChI Key UREBDLICKHMUKA-CXSFZGCWSA-N
Synonym dexamethasone, decadron, dexamethazone, maxidex, decaspray, desametasone, hexadecadrol, dexacort, dexasone, hexadrol
PubChem CID 5743
ChEBI CHEBI:41879
IUPAC Name (1R,2R,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-1-(2-hydroxyacetyl)-2,9a,11a-trimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO

Specifications

Quantity 100 mg
Solubility Information Soluble in water (25mg/mL, clear to slightly hazy, colorless solution)
Formula Weight 392.5g/mol
Physical Form Powder
Recommended Storage Store at Room Temperature (15–30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.