Learn More
Description
Dexamethasone USP is a white to practically white, odorless, crystalline powder. It is stable in air.
Specifications
Specifications
| Chemical Name or Material | Dexamethasone USP (Micronized) |
| Packaging | Jar 60ml, White, HDPE, Round, 33-400, TRIVENI-DB, CT |
| CAS | 50-02-2 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 392.4641 |
| Quantity | 10 g |
| Percent Purity | 97 to 102 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
