Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dexchlorpheniramine Maleate, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602063 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-063 1 kg Amber Glass Bottle
18-602-064 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-602-063 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. DE1271KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

DE127, 2438-32-6, C16H19ClN2·C4H4O4

Spectrum™ Chemical Dexchlorpheniramine Maleate, USP is an antihistamine with anticholinergic properties that is used to treat allergies. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 2438-32-6
Molecular Formula C20H23ClN2O4
Molecular Weight (g/mol) 390.86
InChI Key DBAKFASWICGISY-DASCVMRKSA-N
IUPAC Name (2Z)-but-2-enedioic acid; [(3S)-3-(4-chlorophenyl)-3-(pyridin-2-yl)propyl]dimethylamine
SMILES OC(=O)\C=C/C(O)=O.CN(C)CC[C@@H](C1=CC=C(Cl)C=C1)C1=CC=CC=N1

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 1 kg
UN Number UN2811
Specific Rotation +39.5 to +43.0°
Percent Purity 98 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.