Learn More
Selleck Chemical LLC Dexlansoprazole S4099-50mg
SDP
Supplier:  Selleck Chemical LLC S409950MG
Encompass_Preferred
Dexlansoprazole (T 168390 TAK 390 (R)-Lansoprazole) the dextrorotatory enantiomer of lansoprazole is a proton pump inhibitor (PPI) formulated to have dual delayed-release properties Dexlansoprazole selectively suppresses gastric acid secretion by direct inhibition of the H( ) K( )-ATPase proton pump in the gastric parietal cell
Catalog No. 50-310-8729
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.