Learn More
Abcam Dexmedetomidine Hydrochloride, α² Agonist, Solid, >99% Purity, 10 mg
Description
- Molecular weight: 236.74 g/mol
- Purity: >99%
- Potent, highly selective α2 agonist: Ki values are 1.08 and 1,750 nM at α2 and α1 receptors, respectively
- Active enantiomer: Medetomidine (ab120787)
- Properties: Sedative, analgesic, anxiolytic, and antiulcerative
- Form: Solid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | α2 Adrenoceptor |
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Quantity | 10 mg |
| For Use With (Application) | Sedative, analgesic, anxiolytic, antiulcerative |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.