Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Dexmedetomidine Hydrochloride, Selective α2 Agonist, 50 mg

Catalog No. 502707290
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7290 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7290 Supplier Abcam Supplier No. AB12076750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent and highly selective α2 agonist with >99% purity.

  • Purity: >99%
  • Molecular Weight: 236.74 g/mol
  • Properties: Potent, highly selective α2 agonist; active enantiomer of medetomidine (ab120787)
  • Receptor Affinity: Ki values are 1.08 and 1,750 nM at α2 and α1 receptors, respectively
  • Biological Activities: Displays sedative, analgesic, anxiolytic, and antiulcerative properties
  • Storage: Store at ambient

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule α2 receptors
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Form Solid
Content And Storage Store at ambient temperature.
Quantity 50 mg
For Use With (Application) Sedative, analgesic, anxiolytic, antiulcerative
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.