Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dexmedetomidine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D5062100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5062100MG 100 mg
D506220MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5062100MG Supplier TCI America Supplier No. D5062100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 145108-58-3
Molecular Formula C13H17ClN2
Molecular Weight (g/mol) 236.743
MDL Number MFCD12912677
InChI Key VPNGEIHDPSLNMU-MERQFXBCSA-N
Synonym dexmedetomidine hydrochloride, dexmedetomidine hcl, precedex, 4-s-alpha,2,3-trimethylbenzyl imidazole monohydrochloride, unii-1018wh7f9i, dexmedetomidine hcl precedex, s-5-1-2,3-dimethylphenyl ethyl-1h-imidazole hydrochloride, dexdor, 4-1s-1-2,3-dimethylphenyl ethyl-1h-imidazole hydrochloride
PubChem CID 6918081
ChEBI CHEBI:31472
IUPAC Name 5-[(1S)-1-(2,3-dimethylphenyl)ethyl]-1H-imidazole;hydrochloride
SMILES CC1=C(C(=CC=C1)C(C)C2=CN=CN2)C.Cl

Specifications

Melting Point 158°C
Color White
Quantity 100 mg
Physical Form Crystal-Powder at 20°C
Formula Weight 236.74
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Dexmedetomidine Hydrochloride
RTECSNumber NI5156750
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.