Learn More
TARGETMOL CHEMICALS INC DASATINIB HYDROCHLORIDE 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T22303
Encompass_Preferred
Also available in 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dasatinib hydrochloride (BMS-354825 HCl) (BMS-354825) hydrochloride is a highly potent ATP competitive orally active dual Src/Bcr-Abl inhibitor with potent antitumor activity. The Kis are 16 pM and 30 pM for Src and Bcr-Abl respectively. Dasatinib hydrochloride hydrochloride inhibits Bcr-Abl and Src with IC50s of <1.0 nM and 0.5 nM respectively. Dasatinib hydrochloride hydrochloride also induces Apoptosis and Autophagy. purity: 99%
Catalog No. 50-258-0966
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.