Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextran, MW ca 75,000

Catalog No. AAJ6098922
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6098922 100 g
AAJ6098909 10 g
AAJ6098918 50 g
AAJ6098936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ6098922 Supplier Thermo Scientific Chemicals Supplier No. J6098922
Add to Cart

Dextran, CAS # 9004-54-0, is a glucose polymer. It is mainly used to increase electronegativity and reduce erythrocyte aggregation and platelet adhesiveness (anticoagulant activity). | CAS: 9004-54-0 | (C6H9O5)n(C6H10O5)m | 504.44 g/mol

Dextran, MW ca 75,000 is used as a processing aid, stabilizer, thickener, and surface finishing agent.. Further, it is used in soft center confections. In addition to this, it is used as a barley malt substitute and as a plasma volume expander.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Dextran is a glucose polymer biosynthesized by the non-pathogenic Leuconostoc mesenteroides

Application

• Dextran is a complex branched glucan that can be used to enhance electronegativity and reduce erythrocyte aggregation and platelet adhesiveness (anticoagulant activity)

• This compound promotes the activation of plasminogen by inhibiting α-2 antiplasmin

• Dextran can be used to reduce thrombosis in animal models

• It is used in osmotic stress techniques where osmotic pressure is applied to biological molecules • Dextran can be used as an immobilization agent in biosensors

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-54-0
Molecular Formula (C6H9O5)n(C6H10O5)m
Molecular Weight (g/mol) 504.44
MDL Number MFCD00130935
InChI Key FZWBNHMXJMCXLU-UHFFFAOYNA-N
Synonym dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal
PubChem CID 4125253
IUPAC Name Dextran
SMILES O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O

Specifications

Color White
Physical Form Powder
Merck Index 14,2948
Solubility Information Soluble in water,dimethyl formamide and glycerol.
Quantity 100 g
Formula Weight ≈75 kd
Chemical Name or Material Dextran
EINECSNumber 232-677-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.