Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Dextran Sulfate, Anhydroglucose Polymer, 500,000 g/mol, Solid, 10 g

Catalog No. 502704572
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4572 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4572 Supplier Abcam Supplier No. AB14656910G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Anhydroglucose polymer used for probe hybridization and selective lipoprotein precipitation.

  • Molecular weight: 500,000 g/mol
  • Form: Solid
  • Composition: 100% Dextran sulfate
  • Applications: Selective precipitation of lipoproteins, probe hybridization of membrane-immobilized DNA, release of DNA from DNA-histone complexes, and inhibition of RNA binding to ribosomes
  • Key Benefit: Accelerates the rate of probe hybridization by 10 fold when used as an additive

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% Dextran sulfate
Content And Storage Store at 4°C.
Form Solid
Shipping Condition Ambient
Target Molecule Lipoproteins, DNA, RNA
Formulation 100% Dextran sulfate
Quantity 10 g
For Use With (Application) Selective precipitation of lipoproteins, probe hybridization of membrane-immobilized DNA, release of DNA from DNA-histone complexes, inhibition of RNA binding to ribosomes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.