Learn More
Abcam Dextran Sulfate, Anhydroglucose Polymer, 500,000 g/mol, Solid, 50 g
Description
- Molecular Weight: 500,000 g/mol
- Form: Solid
- Composition: 100% Dextran sulfate
- Applications: Selective precipitation of lipoproteins, probe hybridization of membrane-immobilized DNA, release of DNA from DNA-histone complexes, and inhibition of RNA binding to ribosomes
- Usage: Additive in probe hybridization solutions, accelerating the rate of hybridization by 10 fold
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 100% Dextran sulfate |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Shipping Condition | Ambient |
| Target Molecule | DNA, RNA, Lipoproteins |
| Formulation | 100% Dextran sulfate |
| Quantity | 50 g |
| For Use With (Application) | Selective precipitation of lipoproteins, probe hybridization, release of DNA from DNA-histone complexes, inhibition of RNA binding to ribosomes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.