Learn More
Dextran sulfate sodium salt (400,000-600,000 M.Wt), MP Biomedicals™
Shop All MP Biomedicals, Inc ProductsSpecifications
Specifications
| Color | White |
| Physical Form | Powder |
| Quantity | 10 g |
| IUPAC Name | ethyl 4-aminobenzoate |
| Formula Weight | 400,000 to 600,000 |
| Chemical Name or Material | Dextran Sulfate Sodium Salt |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.