Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextrin, Commercial Grade Type III, MP Biomedicals™

Catalog No. ICN16005790 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN16005790 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN16005790 Supplier MP Biomedicals, Inc Supplier No. 0216005790
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9004-53-9
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.438
InChI Key FYGDTMLNYKFZSV-MRCIVHHJSA-N
Synonym dextrin, dextrins, fortodex, dextrid, corn dextrin, dextrina bianca, british gum, crystal gum, dextrin 3, caloreen
PubChem CID 62698
IUPAC Name (3R,4S,5S,6R)-2-[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O

Specifications

Color White
Quantity 500 g
Physical Form Powder
Chemical Name or Material Dextrin, Commercial Grade Type III
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.