Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Dextrin Sepharose™ High Performance Media
GSA_VA

Catalog No. 45001584 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-584 100 mL
45-001-583 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-001-584 Supplier Cytiva Supplier No. 28935598
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Provide convenient affinity purification of recombinant proteins with maltose binding protein (MBP) tags

  • Physiological conditions and mild elution preserves activity of target protein
  • Robust, stable medium allows narrow peaks in elution of target protein
  • Compatible with commonly used aqueous buffers and easily regenerated with 0.5M NaOH
  • Easy scale-up

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mL
Material Dextrin Sepharose
Particle Size 34 μm
Matrix Affinity Resin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.