Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextromethorphan hydrobromide, Tocris Bioscience™
SDP

Catalog No. 307150 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-715-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 30-715-0 Supplier R&D Systems Supplier No. 3071/50

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

NMDA antagonist

Non-competitive, low affinity NMDA receptor antagonist. Antitussive, anticonvulsive and neuroprotective agent.

Chemical Identifiers

CAS 125-69-9
Molecular Formula C18H28BrNO2
Molecular Weight (g/mol) 370.331
InChI Key STTADZBLEUMJRG-IKNOHUQMSA-N
Synonym dextromethorphan hydrobromide, antussan, demorphan, dormethan, metrorat, medicon, tusilan, tussade, d-methorphan hydrobromide, romilar
PubChem CID 5462351
SMILES CN1CCC23CCCCC2C1CC4=C3C=C(C=C4)OC.O.Br

Specifications

Quantity 50 mg
Formula Weight 352.31
Percent Purity >99%
Chemical Name or Material Dextromethorphan hydrobromide
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.