Learn More
Description
Dextromethorphan Hydrobromide USP is a practically white crystals or crystalline powder, having a faint odor. Slightly hygroscopic.
Alternative name(s): Dextromethorphan hydrobromide monohydrate
Specifications
Specifications
| Chemical Name or Material | Dextromethorphan Hydrobromide USP |
| Packaging | Jar |
| CAS | 6700-34-1 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 370.3282 |
| Quantity | 1 kg |
| Percent Purity | 98 to 102 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
