Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextrose Anhydrous Bioreagent Grade, Vion Biosciences
SCI_ED

Catalog No. S11082 Shop All Aldon Products
Change view
Click to view available options
Quantity:
10 kg
25 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S11082 25 kg
S11074 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S11082 Supplier Aldon Corporation Supplier No. VNGG700025KG
Add to Cart
Add to Cart

Bioreagent grade for molecular biology, DNase-free, Rnase-free, ≥ 99.5%

  • SPECIFICATIONS
  • Appearance (Color): White to Off-White
  • Appearance (Form): Crystalline Powder
  • Purity: ≥ 99.5%
  • Solubility (2g/15 mL): Clear and Colorless
  • Identification (FTIR): Conforms to Structure
  • Conductivity: ≤ 20 μS/cm
  • Specific Rotation: +52.6° to +53.2°
  • Water (KFV): ≤ 1.0%
  • Lead: ≤ 0.5 ppm
  • Sulfates: ≤ 200 ppm
  • Chloride: ≤ 125 ppm
  • Sulfated Ash: ≤ 0.1%
  • Clacium: ≤ 200 ppm
  • Endotoxins ≤ 1.25 EU/g
  • DNase: None Detected
  • RNase: None Detected

Chemical Identifiers

Molecular Weight (g/mol) 180.16 g/mol

Specifications

Boiling Point Decomposes
Quantity 25 kg
Percent Purity ≥ 99.5%
Grade Reagent
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.