Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextrose, Anhydrous, Granular, BiotechGrade, Spectrum™ Chemical
SDP

Catalog No. 18602109 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-109 500 g Poly Bottle
18-602-110 2.5 kg Poly Bottle
18-602-111 12 kg Poly Bottle
18-602-112 45 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-602-109 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D1082500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

D1082, 50-99-7, C6H12O6

Spectrum™ Chemical Dextrose, Anhydrous, Granular, BiotechGrade is a simple monosaccharide sugar and is commonly referred to as glucose. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key WQZGKKKJIJFFOK-UHFFFAOYNA-N
IUPAC Name 6-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMILES OCC1OC(O)C(O)C(O)C1O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 500 g
Grade Biotech
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Dextrose, Anhydrous, Granular
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.