Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextrose, Anhydrous, Purified (D-Glucose), Reagents

Catalog No. 01337292
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-292 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-292 Supplier Reagents Holdings Llc Supplier No. C2195000500A5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dextrose, Anhydrous, Purified (D-Glucose), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key WQZGKKKJIJFFOK-UHFFFAOYNA-N
Synonym d-Glucose
IUPAC Name 6-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMILES OCC1OC(O)C(O)C(O)C1O

Specifications

Boiling Range 233°C
Melting Point 146°C
Color White
pH 5.9
Boiling Point 233°C
Melting Range 146°C
Packaging Poly Bottle
Quantity 500 g
Specific Gravity 1.562
Grade Pure
Physical Form Solid
Chemical Name or Material Dextrose, Anhydrous, Purified (D-Glucose)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.