Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextrose, Monohydrate, Multi-Compendial, U.S.P., J.T. Baker™

Catalog No. 02002711 Shop All Avantor Products
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-002-711 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-002-711 Supplier Avantor Supplier No. 191306
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Multi-compendial grade for manufacturing, processing or repackaging applications.

Chemical Identifiers

CAS 10-1-5996
Molecular Formula C6H14O7
Molecular Weight (g/mol) 198.17
InChI Key OSNSWKAZFASRNG-WNFIKIDCSA-N
Synonym 6-hydroxymethyl oxane-2,3,4,5-tetrol hydrate, d-galactose hydrate, dextrose monohydrate, usp, .alpha.-d-glucose hydrate, dextrose, u.s.p.
PubChem CID 133126654
IUPAC Name (2R,3S,4R,5R,6S)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol;hydrate
SMILES C(C1C(C(C(C(O1)O)O)O)O)O.O

Specifications

Packaging Poly Pail
Quantity 5 kg
Formula Weight 198.17
Percent Purity 97.5 to 102.0%
Grade Multi-Compendial/USP
Chemical Name or Material Dextrose, Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.