Learn More
Honeywell™ DF300 Flat-Fold N95 Respirators
Catalog No. 19087002 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Quantity:
20/Box, 12 Boxes/Cs.
50/Box, 12 Boxes/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-087-002 50/Box, 12 Boxes/Cs.
19-087-003 20/Box, 12 Boxes/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-087-002 Supplier Honeywell™ Supplier No. DF300N95B
Add to Cart
Add to Cart
This item is not returnable. View return policy
NIOSH approved disposable respirators are constructed with a multilayered absorption media that repels moisture and features a smooth inner lining for added comfort.
  • Exclusive design that minimizes the pressure points and fits comfortably on most faces
  • Humidity and moisture resistant filter media that repels moisture for a longer lasting respirator
  • Strong, rigid outer shell prevents masks from collapsing in high humidity
  • Dual welded head straps secure the respirator comfortably
  • Latex free woven head straps reduce the possibility of breakage
Specifications
Aerosol Type N - Not Resistant to Oil
Certifications (NIOSH) NIOSH 42 CFR 84, N95
Design Type Flat-fold
Disposable Single-use
Filter Type N95
Product Type General Purpose Particulate Respirator
Size One Size
With Exhalation Valve No
Filter Efficiency 95%
Latex Free Yes
Model DF300N95B
Nosepiece Type Adjustable Nose Clip
Quantity 50/Box, 12 Boxes/Cs.
Strap Attachment Type Woven Head Straps
FDA Cleared Surgical Mask No