Learn More
Apexbio Technology LLC Anti-SLAMF7/CS1 Antibody (Elotuzumab)(Synonyms: Empliciti, HuLuc63, BMS-901608, Anti-CS1 antibody, Anti-SLAM family member 7 antibody), 1mg.
SDP
Supplier:  Apexbio Technology LLC F14821
Encompass_Preferred
Elotuzumab (CAS 915296-00-3) is a monoclonal antibody directed against the signaling lymphocytic activation molecule family member 7 (SLAMF7 also termed CS1) a surface receptor predominantly expressed on plasma cells including multiple myeloma cells Elotuzumab alone demonstrates limited direct anti-myeloma activity in relapsed or refractory multiple myeloma (RRMM) However when administered in combination with other anti-myeloma agents it enhances therapeutic response and improves patient outcomes This antibody is utilized in biomedical research on plasma cell malignancies and therapeutic combination strategies
Catalog No. 50-000-61336
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.