Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DGAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01670138
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-138 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-138 Supplier Invitrogen™ Supplier No. PA116985
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is not applicable for IHC-paraffin embedded sections. Suggested positive control: HepG2 lysate.

The enzyme encoded by this gene utilizes diacylglycerol and fatty acyl CoA as substrates in order to catalyze the final stage of triacylglycerol synthesis. It is also involved in cellular as well as physiological metabolic processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DGAT1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene DGAT1
Gene Accession No. O75907, Q9ERM3, Q9Z2A7
Gene Alias ACAT related gene product 1; ACAT-related gene product 1; Acyl coenzyme A; acyl coenzyme A:cholesterol acyltransferase related gene 1; acyl-CoA retinol O-fatty-acyltransferase; acyl-CoA:diacylglycerol acyltransferase; acyl-coenzyme A:diacylglycerol acyltransferase 1; AGRP1; ARAT; ARGP1; C75990; D15Ertd23e; Dgat; DGAT1; dgat1a; DGAT1B; diacylglycerol acyltransferase; diacylglycerol acyltransferase 1; diacylglycerol O-acyltransferase 1; diacylglycerol O-acyltransferase 1a; diacylglycerol O-acyltransferase homolog 1; diacylglycerol O-acyltransferase homolog 1a; DIAR7; Diglyceride acyltransferase; EC 2.3.1.20; hypothetical protein LOC325875; putative diacylglycerol acyltransferase 1; retinol O-fatty-acyltransferase; wu:fd36g11; zgc:77691
Gene Symbols DGAT1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region (within residues 200-300) of human DGAT1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13350, 84497, 8694
Target Species Human, Mouse, Rat, Zebrafish
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.