Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGKG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557727
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557727 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557727 Supplier Thermo Scientific Supplier No. PA557727
Only null left
Add to Cart
Add to Cart

DGKG Polyclonal Antibody

DGKG Polyclonal Antibody for ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen DGKG
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DGKG
Gene Accession No. P49619
Gene Alias 2900055E17Rik; 88 kDa diacylglycerol kinase; 90kDa; AI854428; DAG kinase gamma; Dagk3; DGKG; DGK-gamma; Diacylglycerol kinase gamma; diacylglycerol kinase, gamma; diacylglycerol kinase, gamma 3; diacylglycerol kinase, gamma 90kDa; diacylglyerol kinase gamma; diglyceride kinase gamma; E430001K23Rik; mKIAA4131
Gene Symbols DGKG
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DGKG. Recombinant protein control fragment (Product #RP-96269).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1608
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.