Learn More
ATCC DH82; Malignant Histiocytosis; Dog (Canis familiaris)
Supplier:  ATCC CRL3590
Encompass
DH82 is a macrophage-like cell from a 10-year-old male golden retriever with malignant histiocytosis. DH82 can be used in immunology research. This product is an ATCC manufactured and accessioned progeny of ATCC CRL-10389 cited in US Pat. No. 5,192,679.
Catalog No. NC2468594
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.