Learn More
Apexbio Technology LLC Dihydro-β-erythroidine hydrobromide(Synonyms: DHβE hydrobromide, Dihydro-beta-erythroidine hydrobromide, Dihydro-β-erythroidine HBr), 50mg, CAS: 29734-68-7.
SDP
Supplier:  Apexbio Technology LLC B703050
Encompass_Preferred
Dihydro- -erythroidine hydrobromide (CAS 29734-68-7) abbreviated DH E is a competitive antagonist of neuronal nicotinic acetylcholine receptors (nAChRs) Acting directly at nAChR subunits DH E blocks acetylcholine-mediated receptor activation in neurons Its inhibitory potency (IC50) varies by receptor subtype notably 0 37 M for 4 2 0 41 M for 3 2 and 0 19 M for 4 4 nAChRs In vivo studies demonstrate DH E s capacity to cross the blood-brain barrier attenuating nicotine-induced central nervous system responses Due to these antagonist properties DH E is frequently utilized as a research tool in studies investigating nAChR function and neuropharmacology
Catalog No. 50-000-61287
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.