Learn More
Selleck Chemical LLC Sarpogrelate hydrochloride S3706-10mg
SDP
Supplier:  Selleck Chemical LLC S370610MG
Encompass_Preferred
Sarpogrelate (MCI-9042) is a selective 5-HT2A antagonist with pKi values of 8 52 7 43 and 6 57 for 5-HT2A 5-HT2C and 5-HT2B receptors respectively
Catalog No. 50-310-9283
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.