Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHRS4 Antibody [Alexa Fluor Plus 555], R&D Systems™
SDP

Catalog No. RD10038734
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10038734 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10038734 Supplier R&D Systems Supplier No. NBP298675AFP555

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

DHRS4 Polyclonal antibody specifically detects DHRS4 in Human samples. It is validated for Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin)

Specifications

Antigen DHRS4
Applications Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor Plus 555
Formulation 50mM Sodium Borate
Gene Alias CR, dehydrogenase/reductase (SDR family) member 4, dehydrogenase/reductase (SDR family) member 4 like 2A, dehydrogenase/reductase SDR family member 4, EC 1.1.1, EC 1.1.1.184, FLJ11008, humNRDR, NADP-dependent retinol dehydrogenase, NRDRNADPH-dependent retinol dehydrogenase/reductase, Peroxisomal short-chain alcohol dehydrogenase, PHCR, PSCD, SCAD-SRLNADPH-dependent carbonyl reductase/NADP-retinol dehydrogenase, SDR25C1, SDR25C2, SDR-SRL, short chain dehydrogenase/reductase family 25C, member 1, short chain dehydrogenase/reductase family 25C, member 2, Short-chain dehydrogenase/reductase family member 4
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DHRS4 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 10901
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.