Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dhso Crosslinker

Catalog No. 909599100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-959-9100MG 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-959-9100MG Supplier Sigma Aldrich Supplier No. 909599100MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

DHSO Crosslinker is a homobifunctional, acidic residue-targeting, sulfoxide-containing crosslinker for analysis of protein-protein interactions (PPIs) through crosslinking mass spectrometry (XL-MS). DHSO possesses two hydrazide-reactive groups for targeting aspartic acid (Asp) and glutamic acid (Glu) and two symmetrical C-S cleavable bonds adjacent to the central sulfoxide. While former amine-reactive reagents react with lysine (Lys) residues in proximity to the PPI, targeting acidic residues provides further opportunity to characterize protein interaction surfaces at which Asp and Glu are often prevalent. Additionally, the 12.4 +a spacer yields tagged peptides post-cleavage for unambiguous identification by collision-induced dissociation in tandem MS. DHSO Crosslinker provides complementary data to Lys-targeting reagents and will find wide utility in the elucidation of PPIs, study of proteins that function as complexes, quantification of structural dynamics, and quest for targeting undruggable protein targets.Technology Spotlight: Cross-Linkers for Elucidation of Protein-Protein InteractionsDeveloping an Acidic Residue Reactive and Sulfoxide-Containing MS-Cleavable Homobifunctional Cross-Linker for Probing Protein-Protein InteractionsDevelopment of a Novel Sulfoxide-Containing MS-Cleavable Homobifunctional Cysteine-Reactive Cross-Linker for Studying Protein-Protein InteractionsStructural dynamics of the human COP9 signalosome revealed by cross-linking mass spectrometry and integrative modelingSubject to US Patent Application #15/613,065 of the Regents of the University of California

Specifications

Density 1.04746 g/mL
Molecular Formula C15H30O7
Refractive Index n/D 1.4492
Linear Formula C15H30O7
Quantity 1 g
Molecular Weight (g/mol) 322.39 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.