Learn More
DHX32 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
This is a rabbit polyclonal anti-DHX32 antibody raised against part of the C-terminal chain of human DHX32.
Specifications
Specifications
| Antigen | DHX32 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.17 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | DHX32 |
| Gene Accession No. | Q7L7V1, Q8BZS9 |
| Gene Alias | DDX32, DEAD/H box 32, DEAD/H helicase like protein 1, DEAH box protein 32, DHLP1, DHX32, HuDDX32 |
| Gene Symbols | Dhx32 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.