Learn More
AdipoGen Di-(2-picolyl)amine,Chemical. CAS: 1539-42-0
SDP
Supplier:  AdipoGen CDXB0134G005
Encompass_Preferred
Di-(2-picolyl)amine, AdipoGen Life Sciences. Chemical. CAS: 1539-42-0. Formula: C12H13N3. MW: 199.25. Synthetic Building block for synthesis. DPA is a secondary amine with two picolyl substituents. The compound is a tridentate ligand in coordination chemistry and commonly used to produce Zn-based chemosensors/probes, such as Zinpry. As a tridentate ligand this compound provides three nitrogen donors that affords good selectivity for Zn2+ over biologically relevant metals such as Na+, K+, Mg2+ and Ca2+, and leaves coordination sites free for anion binding. The amino nitrogen of the DPA group is a good candidate as an electron donor in either photoinduced electron transfer or photoinduced charge transfer (PET or PCT) sensors. Zn(II)¿DPA complexes are widely used in anion recognition and sensing.
Catalog No. 50-114-6935
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.