Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-2-pyridyl Ketoxime 98.0+%, TCI America™
SDP

Catalog No. D17091G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17091G 1 g
D17095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D17091G Supplier TCI America Supplier No. D17091G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1562-95-4
Molecular Formula C11H9N3O
Molecular Weight (g/mol) 199.213
MDL Number MFCD00006311
InChI Key RZHHBDXWQUIRSQ-LUAWRHEFSA-N
Synonym di-2-pyridyl ketoxime, di-2-pyridyl ketone oxime, bis 2-pyridyl ketone oxime, 2,2'-dipyridyl ketoxime, di-2-pyridylketoxime, methanone, di-2-pyridinyl-, oxime, kloqkbnabuidiu-uhfffaoysa-n, n-bis pyridin-2-yl methylidene hydroxylamine, di 2-pyridinyl methanone oxime #
PubChem CID 5375567
IUPAC Name 2-[(Z)-nitroso(1H-pyridin-2-ylidene)methyl]pyridine
SMILES C1=CC=NC(=C1)C(=C2C=CC=CN2)N=O

Specifications

Melting Point 142°C
Color White-Yellow
Quantity 1 g
Formula Weight 199.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Di-2-pyridyl Ketoxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.