Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

di-Ammonium Hydrogen Phosphate, MilliporeSigma™
GSA_VA

Catalog No. M1012079050 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
50 kg
Packaging:
Fibre carton
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1012079050 50 kg Fibre carton
M1012070500 500 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1012079050 Supplier MilliporeSigma Supplier No. 1012079050
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

For analysis EMSURE™ ACS, Reag. Ph Eur

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-28-0
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
PubChem CID 24540
ChEBI CHEBI:63051
IUPAC Name phosphoric acid diamine
SMILES N.N.OP(O)(O)=O

Specifications

Physical Form Solid
Quantity 50 kg
Solubility Information Soluble in water: 690g/L at 20°C
Formula Weight 132.05
Percent Purity ≥99.0%
Grade ACS
Odor Stinging
Packaging Fibre carton
Density 1.619g/cm3 at 20°C
Chemical Name or Material di-Ammonium hydrogen phosphate
EINECSNumber 231-987-8
Recommended Storage Store at 5° to 30°C (41° to 86°F), Tightly closed. Dry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.