Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

di-Lithium tetraborate, MilliporeSigma™

Catalog No. M1056991000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1056991000 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1056991000 Supplier MilliporeSigma Supplier No. 1056991000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Di-Lithium tetraborate for analytical use with 98.0% minimum assay.

  • Chemical Formula: Li2B4O7
  • Molecular Weight: 169.12
  • Form: Solid
  • Assay: 98.0% (acidimetric)
  • Melting Point: 930°C
  • Density: 2.35 g/cm³ at 20°C
  • Bulk Density: 330 kg/m³
  • Solubility: 141.2 g/L
  • pH: 9.1
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12007-60-2
Molecular Formula B4Li2O7
Molecular Weight (g/mol) 169.11
MDL Number MFCD00011083
InChI Key RDZCISUGRQUANA-UHFFFAOYSA-N
Synonym di-Lithium tetraborate
IUPAC Name dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Specifications

Color White
pH 9.1
Physical Form Solid
Quantity 1 kg
Assay Percent Range 98.00%
Linear Formula Li2B4O7
Solubility Information 5g/L (2°C)
Formula Weight 169.12
Percent Purity 98.0% (acidimetric)
Concentration or Composition (by Analyte or Components) Assay (acidimetric): 98.0%, Ca: 0.005%, Chloride (Cl): 0.005%, Fe: 0.001%, Heavy metals (as Pb): 0.005%, K: 0.005%, Mg: 0.001%, Na: 0.005%, Phosphate (PO4): 0.002%, Sulfate (SO4): 0.005%
Manufacturing Origin Germany
Packaging Plastic Bottle
Product Line Supelco
Appearance White Solid
Specific Gravity 2.35
Density Bulk Density: 330 kg/m3
Assay Acidimetric
Chemical Name or Material Lithium Tetraborate, For Analysis
Show More Show Less
Recommended Storage Store without restrictions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.