Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-n-alkyl Adipate [Plasticizer], TCI America™
SDP

Catalog No. A1386500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1386500ML 500 mL
A138625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1386500ML Supplier TCI America Supplier No. A1386500ML

Chemical Identifiers

MDL Number MFCD00675047
Synonym Adipic Acid Di-n-alkyl Ester

Specifications

Color Yellow
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Di-n-alkyl Adipate [Plasticizer]
EINECSNumber (2)-0861
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.