Learn More
Sigma Organic Chemistry Di-N-Butylammonium Lead(Ii) Tetraiodide N = 1 | 1G | 132069-10-4
Supplier: Sigma Organic Chemistry 9109611G

Di-N-Butylammonium Lead(Ii) Tetraiodide N = 1, 1G
About This Item:
Linear Formula: (C4H9Nh3)2Pbi4
Unspsc Code: 12352101
Storage: Room Temperature
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.