Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-n-octyl phenylphosphonate, 97%

Catalog No. AAA1346814
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1346814 25 g
AAA1346806 5 g
AAA1346822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1346814 Supplier Thermo Scientific Chemicals Supplier No. A1346814
Add to Cart

CAS: 1754-47-8 | C22H39O3P | 382.53 g/mol

Di-n-octyl phenylphosphonate acts as a plasticizer, which is useful in ion-selective electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-octyl phenylphosphonate acts as a plasticizer, which is useful in ion-selective electrodes.

Solubility
Miscible with tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1754-47-8
Molecular Formula C22H39O3P
Molecular Weight (g/mol) 382.53
MDL Number MFCD00009561
InChI Key HAKMAMKAFTZXOZ-UHFFFAOYSA-N
Synonym dioctyl phenylphosphonate, di-n-octyl phenylphosphonate, dioctylphenylphosphonate, phenylphosphonic acid dioctyl ester, benzenephosphonic acid, dioctyl ester, di-n-octyl phenyl phosphonate, dioctyl phenyl phosphonate, phosphonic acid, phenyl-, dioctyl ester, phosphonic acid, p-phenyl-, dioctyl ester, di-n-octylphenylphosphonat
PubChem CID 15641
IUPAC Name dioctoxyphosphorylbenzene
SMILES CCCCCCCCOP(=O)(OCCCCCCCC)C1=CC=CC=C1

Specifications

Density 0.932
Boiling Point 219°C to 220°C (0.7 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4795
Quantity 25 g
Beilstein 2948496
Solubility Information Miscible with tetrahydrofuran.
Formula Weight 382.53
Percent Purity 97%
Chemical Name or Material Di-n-octyl phenylphosphonate
EINECSNumber 217-142-6
RTECSNumber TA0379000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.