Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-n-octyl Phthalate Solution, 100μg/mL in methanol, Ultra Scientific

Catalog No. USPS1501
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-PS-1501 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-PS-1501 Supplier Agilent Technologies Supplier No. PS1501
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 117-84-0 , 67-56-1
Molecular Formula C24H38O4
Molecular Weight (g/mol) 390.564
InChI Key MQIUGAXCHLFZKX-UHFFFAOYSA-N
PubChem CID 8346
ChEBI CHEBI:34679
IUPAC Name dioctyl benzene-1,2-dicarboxylate
SMILES CCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCC

Specifications

CAS Max % 99.99, 0.01
Analyte or Component Names Methanol,di-n-octyl phthalate
Element Methanol, di-n-octyl Phthalate
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material Di-n-octyl Phthalate Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.