Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di(N-succinimidyl) 3,3'-Dithiodipropionate 97.0+%, TCI America™
SDP

Catalog No. D24731G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24731G 1 g
D2473200MG 200 mg
D24735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D24731G Supplier TCI America Supplier No. D24731G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57757-57-0
Molecular Formula C14H16N2O8S2
Molecular Weight (g/mol) 404.408
MDL Number MFCD00042045
InChI Key FXYPGCIGRDZWNR-UHFFFAOYSA-N
PubChem CID 93313
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 3-[[3-(2,5-dioxopyrrolidin-1-yl)oxy-3-oxopropyl]disulfanyl]propanoate
SMILES C1CC(=O)N(C1=O)OC(=O)CCSSCCC(=O)ON2C(=O)CCC2=O

Specifications

Melting Point 133°C
Quantity 1 g
Synonym 3,3′C-Dithiodipropionic Acid Bis(N-succinimidyl Ester), 3,3′C-Dithiodipropionic Acid Di(N-succinimidyl Ester), DTSP
Formula Weight 404.41
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Di(N-succinimidyl) 3,3′-Dithiodipropionate [Cross-linking Reagent]
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.