Learn More
Di(polyethylene Glycol 4-Nonylphenyl) Phosphate (n=approx. 4) (Mono- and Di- Ester mixture), TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Specifications
Specifications
| MDL Number | MFCD00146700 |
| Quantity | 500 g |
| Synonym | Bis(4-nonylphenylpolyoxyethylene) Hydrogen Phosphate, Phosphoric Acid Di(polyethylene Glycol 4-Nonylphenyl) Ester, Polyethylene Glycol Mono-4-nonylphenyl Ether Phosphate |
| Physical Form | Liquid |
| Chemical Name or Material | Di(polyethylene Glycol 4-Nonylphenyl) Phosphate (n=approx. 4) (Mono- and Di- Ester mixture) |
Safety and Handling
| TSCA | No |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.