Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-amyl Dicarbonate 95.0+%, TCI America™
SDP

Catalog No. P135625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P135625G 25 g
P13565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P135625G Supplier TCI America Supplier No. P135625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68835-89-2
Molecular Formula C12H22O5
Molecular Weight (g/mol) 246.30
MDL Number MFCD00038320
InChI Key PGWKKYVDHYLHNF-UHFFFAOYSA-N
Synonym Di-tert-amyl Pyrocarbonate, Pyrocarbonic Acid Di-tert-amyl Ester
PubChem CID 4586057
IUPAC Name bis(2-methylbutan-2-yl) dicarbonate
SMILES CCC(C)(C)OC(=O)OC(=O)OC(C)(C)CC

Specifications

Color Yellow
Boiling Point 71°C
Quantity 25 g
Formula Weight 246.30
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Di-tert-amyl Dicarbonate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.