Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-butyl azodicarboxylate, 97%, Acros Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
870-50-8
C10H18N2O4
230.26
QKSQWQOAUQFORH-VAWYXSNFSA-N
di-tert-butyl azodicarboxylate,di-tert-butylazodicarboxylate,di-tert-butyl diazene-1,2-dicarboxylate,azodicarboxylic acid di-tert-butyl ester,di-tert-butyl azodicarboxylate dbad,pubchem17548,n-tert-butoxycarbonyl imino tert-butoxy formamide,tert-butyl tert-butyl oxycarbonyl diazenyl formate,n-tert-butoxy carbonyl imino tert-butoxy formamide,1,2-diazenedicarboxylic acid, 1,2-bis 1,1-dimethylethyl ester
6034084
tert-butyl (NE)-N-[(2-methylpropan-2-yl)oxycarbonylimino]carbamate
CC(C)(C)OC(=O)N=NC(=O)OC(C)(C)C
870-50-8
C10H18N2O4
230.26
QKSQWQOAUQFORH-VAWYXSNFSA-N
di-tert-butyl azodicarboxylate,di-tert-butylazodicarboxylate,di-tert-butyl diazene-1,2-dicarboxylate,azodicarboxylic acid di-tert-butyl ester,di-tert-butyl azodicarboxylate dbad,pubchem17548,n-tert-butoxycarbonyl imino tert-butoxy formamide,tert-butyl tert-butyl oxycarbonyl diazenyl formate,n-tert-butoxy carbonyl imino tert-butoxy formamide,1,2-diazenedicarboxylic acid, 1,2-bis 1,1-dimethylethyl ester
6034084
tert-butyl (NE)-N-[(2-methylpropan-2-yl)oxycarbonylimino]carbamate
CC(C)(C)OC(=O)N=NC(=O)OC(C)(C)C
C-X Bond Formation (Halogen), Radical initiators
Quantity 25g
Color Yellow
Melting Point 90.0°C to 92.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 96% min. (GC)
Linear Formula (CH3)3CO2NNCO2C(CH3)3
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dichloromethane and other chlorinated,solvents
Formula Weight 230.26
Physical Form Crystals or Crystalline Powder
Percent Purity 97%
Chemical Name or Material Di-tert-butyl azodicarboxylate
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 212-796-9